Learn More
Medchemexpress LLC Fmoc-val-cit-pab-duocarmycin | 99.4% | 1050.59 g·mol-1 | C58H60ClN7O10 | 5 MG
SDP
Catalog No. 5000202341
Encompass_Preferred
Fmoc-Val-Cit-PAB-duocarmycin is a drug-linker conjugate intended for antibody-drug conjugate (ADC) synthesis. It couples a duocarmycin-based cytotoxic payload to an Fmoc-Val-Cit-PAB cleavable spacer, providing a reagent for linker-payload incorporation during ADC development.
  • Cleavable val-cit-pab spacer for protease-triggered release.
  • Duocarmycin-based cytotoxic payload for potent cell killing.
  • High purity (99.4%) with characterized molecular weight.
  • Supplied as a solid, white to yellow, easy-to-handle material.
  • Storage: powder -20°C; in solvent -80°C for long-term stability.
Catalog No. 50-002-02341 Supplier Medchemexpress LLC Supplier No. HY1265325MG
Fmoc-Val-Cit-PAB-duocarmycin is a drug-linker conjugate intended for antibody-drug conjugate (ADC) synthesis. It couples a duocarmycin-based cytotoxic payload to an Fmoc-Val-Cit-PAB cleavable spacer, providing a reagent for linker-payload incorporation during ADC development.
  • Cleavable val-cit-pab spacer for protease-triggered release.
  • Duocarmycin-based cytotoxic payload for potent cell killing.
  • High purity (99.4%) with characterized molecular weight.
  • Supplied as a solid, white to yellow, easy-to-handle material.
  • Storage: powder -20°C; in solvent -80°C for long-term stability.