Learn More
Medchemexpress LLC Fmoc-Val-Cit-PAB-MMAE | 1350456-56-2 | 98.9% | 1345.67 | C73H104N10O14 | 50 MG
SDP
Catalog No. 5000202754
Encompass_Preferred
Fmoc-Val-Cit-PAB-MMAE is an antibody-drug conjugate linker-payload conjugate that couples an Fmoc-protected Val-Cit-PAB cleavable linker to monomethyl auristatin E (MMAE) for use in antibody-drug conjugate research and development.
  • Contains a cleavable Val-Cit-PAB linker that is susceptible to proteolytic release.
  • Delivers monomethyl auristatin E (MMAE), a potent tubulin inhibitor.
  • High reported purity suitable for research applications.
  • Supplied as a white solid and available in multiple pack sizes.
  • Provided with supporting documents such as SDS and certificate of analysis for safe handling and quality verification.
Catalog No. 50-002-02754 Supplier Medchemexpress LLC Supplier No. HY1981150MG
Fmoc-Val-Cit-PAB-MMAE is an antibody-drug conjugate linker-payload conjugate that couples an Fmoc-protected Val-Cit-PAB cleavable linker to monomethyl auristatin E (MMAE) for use in antibody-drug conjugate research and development.
  • Contains a cleavable Val-Cit-PAB linker that is susceptible to proteolytic release.
  • Delivers monomethyl auristatin E (MMAE), a potent tubulin inhibitor.
  • High reported purity suitable for research applications.
  • Supplied as a white solid and available in multiple pack sizes.
  • Provided with supporting documents such as SDS and certificate of analysis for safe handling and quality verification.