Learn More
Medchemexpress LLC HY-41189 250mg , Fmoc-Val-Cit-PAB-PNP CAS:863971-53-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY41189250MG
Encompass_Preferred
HY-41189 250mg Fmoc-Val-Cit-PAB-PNP CAS: 863971-53-3 Fmoc-Val-Cit-PAB-PNP is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1069