Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Val-OH, Advanced ChemTech

Catalog No. 01339945
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Catalog No. Quantity
01-339-945 1 g
01-339-943 5 g
01-339-944 25 g
01-339-946 100 g
4 options

Catalog No. 01-339-945

Supplier: Advanced Chem Tech FV25751G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68858-20-8
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00037124
InChI Key UGNIYGNGCNXHTR-SFHVURJKSA-N
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Val-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.