Learn More
Medchemexpress LLC Fmoc-VAP-MMAE | 1912408-91-3 | 1259.57 | 10 MG
SDP
Catalog No. 5000365446
Encompass_Preferred
Fmoc-VAP-MMAE is an agent-linker conjugate for Antibody-Drug Conjugates (ADC). It consists of an ADC linker, a tubulin inhibitor MMAE (HY-15162), and a protective Fmoc group. It is intended for research use only and is not sold to patients. The appearance is a solid, white to off-white in color.
  • Agent-linker conjugate for ADC
  • Composed of an ADC linker and MMAE, a tubulin inhibitor
  • Features a protective Fmoc group
  • Intended for research use only
  • Appearance: solid, white to off-white in color
Catalog No. 50-003-65446 Supplier Medchemexpress LLC Supplier No. 10MGHY148339
Fmoc-VAP-MMAE is an agent-linker conjugate for Antibody-Drug Conjugates (ADC). It consists of an ADC linker, a tubulin inhibitor MMAE (HY-15162), and a protective Fmoc group. It is intended for research use only and is not sold to patients. The appearance is a solid, white to off-white in color.
  • Agent-linker conjugate for ADC
  • Composed of an ADC linker and MMAE, a tubulin inhibitor
  • Features a protective Fmoc group
  • Intended for research use only
  • Appearance: solid, white to off-white in color