Learn More
Cayman Chemical Fmoc-L-Leucine, Purity: ≥98%, CAS: 35661-60-0, 25 g
SDP
Catalog No. 502891122
Encompass_Preferred
A partial agonist of PPARγ; activates PPARγ with a lower potency (Ki = 15 versus 0.035 µM) but a similar maximal efficacy compared to rosiglitazone; improves insulin resistance in normal, diet-induced glucose-intolerant, and in diabetic db/db mice, yet reduced adipogenic activity
Catalog No. 50-289-1122 Supplier Cayman Chemical Supplier No. 1000488825 G
A partial agonist of PPARγ; activates PPARγ with a lower potency (Ki = 15 versus 0.035 µM) but a similar maximal efficacy compared to rosiglitazone; improves insulin resistance in normal, diet-induced glucose-intolerant, and in diabetic db/db mice, yet reduced adipogenic activity