Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical FmocNH-PEG8-CH2CH2COOH | 100 g | CAS 756526-02-0 | MDL MFCD11041143

Catalog No. 502360234
Encompass

FmocNH-PEG8-CH2CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 756526-02-0
- MDL: MFCD11041143
- InChIKey: VYXGUCLTEWCVRY-UHFFFAOYSA-N
- Molecular Weight: 663.761
- Molecular Formula: C34H49NO12
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid
- SMILES: O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)O

Catalog No. 50-236-0234 Supplier STA Pharmaceutical Supplier No. STAAA11213100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FmocNH-PEG8-CH2CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 756526-02-0
- MDL: MFCD11041143
- InChIKey: VYXGUCLTEWCVRY-UHFFFAOYSA-N
- Molecular Weight: 663.761
- Molecular Formula: C34H49NO12
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid
- SMILES: O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.