Learn More
CPC Scientific H-Phe-Met-Arg-Phe-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific FMRF001B
Encompass
SEQUENCE: H-Phe-Met-Arg-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: FMRFMOLECULAR FORMULA: C29H41N7O5S1MOLECULAR WEIGHT: 599.74STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [74012-06-9]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:SKU(s): FMRF-001A, FMRF-001B
Catalog No. 50-210-9301