Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fms-like tyrosine kinase 3 (FLT3) protein, human, recombinant | >95.0% | 5 UG
SDP

Catalog No. 5000292910
Encompass_Preferred

Recombinant human FMS-like tyrosine kinase 3 (FLT3) extracellular domain fused to a human Fc tag, produced in HEK293 cells. Supplied lyophilized for convenient storage and reconstitution, the protein is intended for binding studies, antibody development, and biochemical assays targeting FLT3 signaling.

  • Recombinant FLT3 extracellular domain with C-terminal human Fc tag.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • Amino acid range N27-S543 (extracellular region).
  • Molecular weight approximately 100-145 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation; store frozen and aliquot after reconstitution.

Catalog No. 50-002-92910 Supplier Medchemexpress LLC Supplier No. HYP751845UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human FMS-like tyrosine kinase 3 (FLT3) extracellular domain fused to a human Fc tag, produced in HEK293 cells. Supplied lyophilized for convenient storage and reconstitution, the protein is intended for binding studies, antibody development, and biochemical assays targeting FLT3 signaling.

  • Recombinant FLT3 extracellular domain with C-terminal human Fc tag.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • Amino acid range N27-S543 (extracellular region).
  • Molecular weight approximately 100-145 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation; store frozen and aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.