Learn More
Eurofins Discoverx Fms (Y969C), active, 250 µg. (14-820M)
Supplier:  Eurofins Discoverx 14820M
Encompass
N-terminal 6xHis-tagged recombinant, human Fms amino acids 538-end containing the mutation Y969C. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9374