Learn More
Selleck Chemical LLC FN-1501 E2658-5MG
SDP
Catalog No. 5000039947
Encompass_Preferred
FN-1501 is a potent inhibitor of Fms-like receptor tyrosine kinase 3 (FLT3) and cyclin-dependent kinase (CDK) with IC50s of 2 47 0 85 1 96 and 0 28 nM for CDK2/cyclin A CDK4/cyclin D1 CDK6/cyclin D1 and FLT3 respectively
Catalog No. 50-000-39947 Supplier Selleck Chemical LLC Supplier No. E26585MG
FN-1501 is a potent inhibitor of Fms-like receptor tyrosine kinase 3 (FLT3) and cyclin-dependent kinase (CDK) with IC50s of 2 47 0 85 1 96 and 0 28 nM for CDK2/cyclin A CDK4/cyclin D1 CDK6/cyclin D1 and FLT3 respectively