Learn More
TARGETMOL CHEMICALS INC FN-1501 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T153355MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FN-1501 is a potent FLT3 and CDK inhibitor (IC50s 2.47 0.85 1.96 and 0.28 nM for CDK2/cyclin A CDK4/cyclin D1 CDK6/cyclin D1 and FLT3 respectively). FN-1501 also has anticancer activity. purity: 96%
Catalog No. 50-258-1231