Learn More
Medchemexpress LLC Fn-439 TFA | 124168-73-6 | MFCD00237459 | 99.8% | 604.58 g·mol⁻¹ | C25H35F3N6O8 | 5 MG
SDP
Catalog No. 5000215603
Encompass_Preferred
FN-439 TFA is the trifluoroacetic acid salt of FN-439, a selective small-molecule inhibitor of collagenase-1 (matrix metalloproteinase). It inhibits collagenase-1 with an IC50 of 1 μM and is supplied as a research reagent for in vitro studies of cancer and inflammatory processes.
  • Selective collagenase-1 inhibitor with reported IC50 of 1 μM.
  • High purity (99.81%) suitable for biochemical assays.
  • Trifluoroacetic acid salt form to aid solubility in aqueous systems.
  • Supplied in research-scale quantities (e.g., five mg) for assay development.
  • Intended for in vitro research applications; not for human use.
Catalog No. 50-002-15603 Supplier Medchemexpress LLC Supplier No. HY100210A5MG
FN-439 TFA is the trifluoroacetic acid salt of FN-439, a selective small-molecule inhibitor of collagenase-1 (matrix metalloproteinase). It inhibits collagenase-1 with an IC50 of 1 μM and is supplied as a research reagent for in vitro studies of cancer and inflammatory processes.
  • Selective collagenase-1 inhibitor with reported IC50 of 1 μM.
  • High purity (99.81%) suitable for biochemical assays.
  • Trifluoroacetic acid salt form to aid solubility in aqueous systems.
  • Supplied in research-scale quantities (e.g., five mg) for assay development.
  • Intended for in vitro research applications; not for human use.