Learn More
Enzo Life Sciences Fibronectin (cellular) (human) monoclonal antibody (DH1) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLFG60100100
Encompass_Preferred
Clone: DH1. Isotype: IgG1. Immunogen: Purified fibronectin from A8387 fibrosarcoma cells. Species reactivity: Human. Specificity: Recognizes the extra domain A (ED) of cellular fibronectin (cFn). Recognizes cFN in malignant tumors, does not cross-react with benign tumors or normal tissues. Applications: ICC, WB. Purity: Protein A affinity chromatography. Formulation: Liquid. In PBS containing 0.1% sodium azide and 1mg/mL BSA.
Catalog No. 50-201-0822