Learn More
AdipoGen anti-Fibronectin (EDA), mAb (blocking) (IST-9) (preservative free)
SDP
Supplier:  AdipoGen AG20B6001YPFC100
Encompass_Preferred
anti-Fibronectin (EDA), mAb (blocking) (IST-9) (preservative free), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes an epitope located in the EDA sequence of human cellular fibronectin (FN). Does not detect extracellular FN (plasma FN). Cross-reacts with mouse, rat, monkey, pig, dog, cow and chicken fibronectin EDA (according to literature). Isotype: Mouse IgG1. Clone: IST-9. Applications: ELISA, FUNC (Blocking), ICC, IHC, WB. Liquid. In 20mM sodium phosphate buffer, pH7.6 containing 150mM sodium chloride.
Catalog No. 50-117-5250