Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2335MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: HFN7.1. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: SW156 cells or Kidney. Cellular Staining: Connective tissue matrix. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 220kDa (monomer); 440kDa (dimer).Chromosome: 2q35. Gene Symbol: FN1. Gene ID: 2335. Ensemble: ENSG00000115414. Uniprot: NP_001293061.
Catalog No. 50-188-4696
Edge