Learn More
Enzo Life Sciences Fn14 (mouse) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522036C050
Encompass_Preferred
Alternative name: TWEAK-R, TNFRSF 12A, Fibroblast growth factor inducible immediate early response protein 14, CD266. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse Fn14 (aa 1-75) is fused to the Fc portion of human IgG1. Specificity: Binds human and mouse TWEAK. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9383