Learn More
Aviva Systems Biology Corp Fnbp1 Antibody - N-terminal region (ARP40800_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fnbp1. It was validated on Western Blot. . Alternate names: 1110057E06Rik, 2210010H06Rik, FBP1, Fbp17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-893