Learn More
ATHENA ES FNC COATING MIX HUMAN 100ML
Catalog No. NC2000952
Encompass_Preferred
Human FNC Mix is a specially formulated serum-free tissue culture reagent containing fibronectin, collagen and albumin that is used to enhance the attachment of adherent cells to plastic flasks or microplates. The unique formula creates an extracellular matrix that dramatically increases the rate of cell attachment to any plastic substratum. The matrix accelerates monolayer formation, especially when fastidious cell types such as human prostrate and breast epithelial cells are being propagated. Mammalian cell cultures will attach and grow more effectively on FNC-coated plastic surfaces when cultured in the appropriate serum-free medium. AthenaES offers several different serum-free media that when used in conjunction with Human FNC Mix, increase the growth of cell cultures dramatically.
Catalog No. NC2000952 Supplier ATHENA ES Supplier No. 0407H
Human FNC Mix is a specially formulated serum-free tissue culture reagent containing fibronectin, collagen and albumin that is used to enhance the attachment of adherent cells to plastic flasks or microplates. The unique formula creates an extracellular matrix that dramatically increases the rate of cell attachment to any plastic substratum. The matrix accelerates monolayer formation, especially when fastidious cell types such as human prostrate and breast epithelial cells are being propagated. Mammalian cell cultures will attach and grow more effectively on FNC-coated plastic surfaces when cultured in the appropriate serum-free medium. AthenaES offers several different serum-free media that when used in conjunction with Human FNC Mix, increase the growth of cell cultures dramatically.