Learn More
Medchemexpress LLC FNC-TP | 2457357-99-0 | 99.9% | 526.16 g/mol | C9H14FN6O13P3 | 5 MG
SDP
Numéro de catalogue. 5000221398
Encompass_Preferred
FNC-TP is the intracellular triphosphate (active) form of a nucleoside reverse transcriptase inhibitor supplied for research applications. It is intended for use in biochemical and cellular assays to study antiviral activity and enzyme interactions, with storage recommendations to preserve stability.
  • Active triphosphate form of a nucleoside reverse transcriptase inhibitor.
  • Antiviral activity against HIV, HBV, and HCV.
  • High purity (>99.9%) suitable for sensitive assays.
  • Available in small-mass vials for biochemical and cellular experiments.
  • Store frozen and protected from light to maintain stability.
Numéro de catalogue. 50-002-21398 Fournisseur Medchemexpress LLC Code fournisseur HY1392625MG
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
FNC-TP is the intracellular triphosphate (active) form of a nucleoside reverse transcriptase inhibitor supplied for research applications. It is intended for use in biochemical and cellular assays to study antiviral activity and enzyme interactions, with storage recommendations to preserve stability.
  • Active triphosphate form of a nucleoside reverse transcriptase inhibitor.
  • Antiviral activity against HIV, HBV, and HCV.
  • High purity (>99.9%) suitable for sensitive assays.
  • Available in small-mass vials for biochemical and cellular experiments.
  • Store frozen and protected from light to maintain stability.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.