Learn More
Medchemexpress LLC FNC-TP | 2457357-99-0 | 99.92% | 526.16 | 1 MG
SDP
Catalog No. 5000347775
Encompass_Preferred
FNC-TP is the intracellular active form of FNC, functioning as a potent nucleoside reverse transcriptase inhibitor (NRTI) with antiviral activity against HIV, HBV, and HCV. Additionally, it serves as a click chemistry reagent containing an azide group, enabling both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Potent nucleoside reverse transcriptase inhibitor (NRTI)
  • Exhibits antiviral activity against HIV, HBV, and HCV
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Intracellularly active form of FNC
  • Accumulates in HIV-1 target cells
Catalog No. 50-003-47775 Supplier Medchemexpress LLC Supplier No. HY1392621MG
FNC-TP is the intracellular active form of FNC, functioning as a potent nucleoside reverse transcriptase inhibitor (NRTI) with antiviral activity against HIV, HBV, and HCV. Additionally, it serves as a click chemistry reagent containing an azide group, enabling both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Potent nucleoside reverse transcriptase inhibitor (NRTI)
  • Exhibits antiviral activity against HIV, HBV, and HCV
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Intracellularly active form of FNC
  • Accumulates in HIV-1 target cells