Learn More
AdipoGen FNDC4 human ELISA Kit
SDP
Supplier:  AdipoGen AG45B0028KI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects human FNDC4 in serum, plasma and cell culture supernatant. It cross-reacts with mouse, rat, monkey and dog FNDC4. It does not cross-react with human, mouse or rat FNDC5 / Irisin. Range 0.078 to 5ng/ml. Sensitivity 40pg/ml. Samples Cell Culture Supernatant, Plasma, Serum. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone irisin. Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure. FNDC4 is an ortholog of FNDC5 with 50% identity and 86% similarity compared to Irisin. FNDC4 as well as FNDC5 are extremely well conserved between species. The human FNDC4 gene is highly enriched in liver, brain tissue and adipocytes.
Catalog No. 50-256-2731