Learn More
Aviva Systems Biology Corp FNDC8 Antibody - N-terminal region (ARP56975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FNDC8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FNDC8. Alternate names: DKFZp434H2215ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-286-247