Learn More
Cayman Chemical FngchInolIn 100mg
SDP
Catalog No. 502937566
Encompass_Preferred
An alkaloid; inhibits sheep COX activity and decreases IL-6-induced proliferation of MH60 mouse hybridoma cells (IC50s = 129 and 3.7 µM, respectively); reduces cyanide-induced increases in secreted glutamate levels and cell death in primary neonatal rat cerebellar granule neurons at 1 and 10 µM; decreases proliferation of PC3 human prostate cancer cells by 63 and 86% at 20 and 30 µM, respectively; reduces tumor cell growth in a PC3 mouse xenograft model at 5 mg/kg per day for 12 days
Catalog No. 50-293-7566 Supplier Cayman Chemical Supplier No. 29243100 MG
An alkaloid; inhibits sheep COX activity and decreases IL-6-induced proliferation of MH60 mouse hybridoma cells (IC50s = 129 and 3.7 µM, respectively); reduces cyanide-induced increases in secreted glutamate levels and cell death in primary neonatal rat cerebellar granule neurons at 1 and 10 µM; decreases proliferation of PC3 human prostate cancer cells by 63 and 86% at 20 and 30 µM, respectively; reduces tumor cell growth in a PC3 mouse xenograft model at 5 mg/kg per day for 12 days