Learn More
Cayman Chemical Fingolimod-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1346747-38-3, 1 mg
SDP
Catalog No. 502934584
Encompass_Preferred
An internal standard for the quantification of fingolimod by GC- or LC-MS
Catalog No. 50-293-4584 Supplier Cayman Chemical Supplier No. 250371 MG
An internal standard for the quantification of fingolimod by GC- or LC-MS