Learn More
Cayman Chemical FngolImod-d4 500ug
SDP
Catalog No. 502934854
Encompass_Preferred
An internal standard for the quantification of fingolimod by GC- or LC-MS
Catalog No. 50-293-4854 Supplier Cayman Chemical Supplier No. 25037500 UG
An internal standard for the quantification of fingolimod by GC- or LC-MS