Learn More
IDEX Health & Science Upchurch FNGRTGHT III PEEK 10-32 10/PK
Supplier:  IDEX Health & Science Upchurch F300X
Encompass_Preferred
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned, Undesignated, Two Piece Fingertight Double Winged Head Nut; 10 to 32 Coned, For Use With: For 0.06 in. O.D. Tubing, Includes: Ferrule.
Catalog No. NC1919057