Learn More
Cayman Chemical FNIII14 (sodium salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502944237
Encompass_Preferred
A peptide inhibitor of cell adhesion; inhibits the adhesion of A375-SM cells to fibronectin-coated plates at 200 µg/ml; inhibits the binding of an antibody targeting the active conformation of β1 integrin on K562 cells and induces the differentiation of ST 13 fibroblasts into adipocytes at 100 µg/ml; decreases atherosclerotic plaque formation in Ldlr-/- mice fed a Western diet; reduces metastatic tumor burden in a 4T1 murine mammary model of metastasis in combination with doxorubicin
Catalog No. 50-294-4237 Supplier Cayman Chemical Supplier No. 4139610 MG
A peptide inhibitor of cell adhesion; inhibits the adhesion of A375-SM cells to fibronectin-coated plates at 200 µg/ml; inhibits the binding of an antibody targeting the active conformation of β1 integrin on K562 cells and induces the differentiation of ST 13 fibroblasts into adipocytes at 100 µg/ml; decreases atherosclerotic plaque formation in Ldlr-/- mice fed a Western diet; reduces metastatic tumor burden in a 4T1 murine mammary model of metastasis in combination with doxorubicin