Learn More
Chemglass Life Sciences Funnel, Standard Stem, 32.8mL, 55mm Top ID
SDP
Catalog No. 50872155
Encompass_Preferred
Funnel, Standard Stem, 32.8mL, 55mm Top ID. Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock. The parallel stem measures 8mm OD. Steam autoclavable at 121°C (250°F).
Catalog No. 50-872-155 Supplier Chemglass Life Sciences Supplier No. CG177001
Funnel, Standard Stem, 32.8mL, 55mm Top ID. Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock. The parallel stem measures 8mm OD. Steam autoclavable at 121°C (250°F).