Learn More
Chemglass Life Sciences Funnel, Weighing, Borosilicate, 20mL Solid Capacity, 10mL Liquid Capacity
SDP
Catalog No. 50872130
Encompass_Preferred
Funnel, Weighing, Borosilicate, 20mL Solid Capacity, 10mL Liquid Capacity.• Available in anti-static PP, PP and borosilicate glass• With flat bottoms and designed to fit narrow neck vessels
Catalog No. 50-872-130 Supplier Chemglass Life Sciences Supplier No. CG176028
Funnel, Weighing, Borosilicate, 20mL Solid Capacity, 10mL Liquid Capacity.• Available in anti-static PP, PP and borosilicate glass• With flat bottoms and designed to fit narrow neck vessels