Learn More
Chemglass Life Sciences Funnel, Standard Stem, 91.8mL, 75mm Top ID
SDP
Catalog No. 50872157
Encompass_Preferred
Funnel, Standard Stem, 91.8mL, 75mm Top ID. Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock. The parallel stem measures 8mm OD. Steam autoclavable at 121°C (250°F).
Catalog No. 50-872-157 Supplier Chemglass Life Sciences Supplier No. CG177003
Add to Cart
Add to Cart
Funnel, Standard Stem, 91.8mL, 75mm Top ID. Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock. The parallel stem measures 8mm OD. Steam autoclavable at 121°C (250°F).