Learn More
Aviva Systems Biology Corp FNTA Antibody - C-terminal region (ARP48521_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48521_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FNTA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FNTA. Alternate names: FPTA, MGC99680, PGGT1A, PTAR2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-59848