Learn More
Aviva Systems Biology Corp FNTA Antibody - N-terminal region (ARP48520_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48520_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FNTA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FNTA. Alternate names: FPTA, MGC99680, PGGT1A, PTAR2ApplicationWB, IHC. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59847