Learn More
New England Biolabs, Inc. Fnu4HI – 1000 units
SDP
Catalog No. 50812094
Encompass
A restriction endonuclease that recognizes the sequence GC^N_GC.
Catalog No. 50-812-094 Supplier New England Biolabs, Inc. Supplier No. R0178L
A restriction endonuclease that recognizes the sequence GC^N_GC.