Learn More
New England Biolabs, Inc. Fnu4HI – 200 units
SDP
Catalog No. 50812095
Encompass
A restriction endonuclease that recognizes the sequence GC^N_GC.
Catalog No. 50-812-095 Supplier New England Biolabs, Inc. Supplier No. R0178S
A restriction endonuclease that recognizes the sequence GC^N_GC.