Learn More
Ran Biotechnologies Inc 10kg of FO-001 fluorinated oil
SDP
Catalog No. NC3660271
Encompass_Preferred
10kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660271 Supplier Ran Biotechnologies Inc Supplier No. FO00110000G
Add to Cart
Add to Cart
Request a Quote
10kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)