Learn More
Ran Biotechnologies Inc 1kg of FO-001 fluorinated oil
SDP
Catalog No. NC3660265
Encompass_Preferred
1kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660265 Supplier Ran Biotechnologies Inc Supplier No. FO0011000G
1kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)