Learn More
Ran Biotechnologies Inc 100g of FO-001 fluorinated oil
SDP
Catalog No. NC3660254
Encompass_Preferred
100g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660254 Supplier Ran Biotechnologies Inc Supplier No. FO001100G
100g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)