Learn More
Ran Biotechnologies Inc 1.5kg of FO-001 fluorinated oil
SDP
Catalog No. NC3660267
Encompass_Preferred
1.5kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660267 Supplier Ran Biotechnologies Inc Supplier No. FO0011500G
1.5kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)