Learn More
Ran Biotechnologies Inc 200g of FO-001 fluorinated oil
SDP
Catalog No. NC3660257
Encompass_Preferred
200g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660257 Supplier Ran Biotechnologies Inc Supplier No. FO001200G
200g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)