Learn More
Ran Biotechnologies Inc 2.5kg of FO-001 fluorinated oil
SDP
Catalog No. NC3660269
Encompass_Preferred
2.5kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660269 Supplier Ran Biotechnologies Inc Supplier No. FO0012500G
2.5kg of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)