Learn More
Ran Biotechnologies Inc 300g of FO-001 fluorinated oil
SDP
Catalog No. NC3660260
Encompass_Preferred
300g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660260 Supplier Ran Biotechnologies Inc Supplier No. FO001300G
300g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)