Learn More
Ran Biotechnologies Inc 500g of FO-001 fluorinated oil
SDP
Catalog No. NC3660264
Encompass_Preferred
500g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660264 Supplier Ran Biotechnologies Inc Supplier No. FO001500G
500g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)