Learn More
Ran Biotechnologies Inc 50g of FO-001 fluorinated oil
SDP
Catalog No. NC3660250
Encompass_Preferred
50g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)
Catalog No. NC3660250 Supplier Ran Biotechnologies Inc Supplier No. FO00150G
50g of FO-001 fluorinated oil (bp 62degC; d1.52; viscosity 0.48cSt)