Learn More
Ran Biotechnologies Inc 10kg of FO-101 fluorinated oil
SDP
Catalog No. NC3660291
Encompass_Preferred
10kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660291 Supplier Ran Biotechnologies Inc Supplier No. FO10110000G
Add to Cart
Add to Cart
Request a Quote
10kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)