Learn More
Ran Biotechnologies Inc 1kg of FO-101 fluorinated oil
SDP
Catalog No. NC3660287
Encompass_Preferred
1kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660287 Supplier Ran Biotechnologies Inc Supplier No. FO1011000G
1kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)