Learn More
Ran Biotechnologies Inc 100g of FO-101 fluorinated oil
SDP
Catalog No. NC3660278
Encompass_Preferred
100g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660278 Supplier Ran Biotechnologies Inc Supplier No. FO101100G
100g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)