Learn More
Ran Biotechnologies Inc 200g of FO-101 fluorinated oil
SDP
Catalog No. NC3660281
Encompass_Preferred
200g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660281 Supplier Ran Biotechnologies Inc Supplier No. FO101200G
200g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)