Learn More
Ran Biotechnologies Inc 2.5kg of FO-101 fluorinated oil
SDP
Catalog No. NC3660289
Encompass_Preferred
2.5kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660289 Supplier Ran Biotechnologies Inc Supplier No. FO1012500G
2.5kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)