Learn More
Ran Biotechnologies Inc 500g of FO-101 fluorinated oil
SDP
Catalog No. NC3660286
Encompass_Preferred
500g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660286 Supplier Ran Biotechnologies Inc Supplier No. FO101500G
500g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)